Would you like to log in again?
TBARS Fluorometric Assay, Eagle Biosciences
Supplier: Eagle Biosciences
TBF39-K01
102995-016EA 451.15 USD InStock
102995-016
TBARS Fluorometric Assay, Eagle Biosciences
Assays ELISAs
2-Thiobarbituric Acid Reactive Substances Fluorometric detects TBARS in serum, plasma, or urine.
  • Dynamic Range: 0.5 - 20 µM
  • Sensitivity: 0.5 µM
  • Incubation: 30 minutes
2-ThioBarbituric Acid Reactive Substances (TBARS) are naturally present in biological specimens and include lipid hydroperoxides and aldehydes which increase in concentration as a response to oxidative stress.1,2 TBARS assay values are usually reported in malonaldehyde (malondialdehyde, MDA) equivalents, a compound that results from the decomposition of polyunsaturated fatty acid lipid peroxides. The TBARS assay is a well-recognized, established method for quantifying these lipid peroxides, although it has been criticized for its reactivity towards other compounds other than MDA.3 This TBARS Cuvette Assay Kit offers the researcher a straightforward, reproducible and consistent method for analyzing urine for lipid peroxidation products.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty