Would you like to log in again?
Tg (Thyroglobulin) Ultrasensitive ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
4018
102995-024EA 961.15 USD InStock
102995-024
Tg (Thyroglobulin) Ultrasensitive ELISA, Eagle Biosciences
Assays ELISAs
Tg (Thyroglobulin) Ultra ELISA is used to determine human thyroglobulin (hTg) in serum.
  • Dynamic Range: 0.03 - 3 ng/mL
  • Sensitivity: 0.03 ng/mL
  • Incubation: overnight
Biochemically, Tg is to be understood as a rather complex family of molecules. It is micro heterogeneous with inter- and intra-individual variations (iodination degree, carbohydrate contents etc.). Dimers and several fragments also exist. Additional heterogeneity is due to malignant de-differentiation. Specifically and unspecifically interfering factors in individual sera cause further problems. Therefore, the Tg determination still represents a rather ambitious method.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty