Would you like to log in again?
Mouse tPA activity ELISA Kit, Eagle Biosciences
Supplier: Eagle Biosciences
TPA19-K01
102995-040EA 1477.69 USD InStock
102995-040
Mouse tPA activity ELISA Kit, Eagle Biosciences
Assays ELISAs
The Mouse tPA Activity ELISA determines active tPA in mouse biological fluids.
  • Dynamic Range: 0.1 - 50 ng/ml
  • Sensitivity: 0.1 ng/ml
  • Incubation: 2.5 hours
Tissue-type Plasminogen Activator (tPA) is a member of the serine proteinase family. tPA functions to lyse fibrin clots into soluble plasmin fragments. tPA is active in two forms, single chain and two-chain. The two-chain tPA is created via interaction with the plasmin product cleaving the single chain. This two-chain form is regarded as the more active form.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty