Would you like to log in again?
Human tPA Activity ELISA Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
TPA39-K01
102995-044EA 1412.31 USD InStock
102995-044
Human tPA Activity ELISA Kit, Eagle Biosciences, Inc.
Assays ELISAs
Human Tissue-type Plasminogen Activator (tPA) Activity ELISA determines tPA in biological fluids.
  • Dynamic Range: 0.1 - 5 ng/ml
  • Sensitivity: 0.006 IU/µL
  • Incubation: 2.5 hours
Tissue-type Plasminogen Activator (tPA) is a member of the serine proteinase family. tPA functions to lyse fibrin clots into soluble plasmin fragments. tPA is active in two forms, single chain and two chain. The two-chain tPA is created via interaction with the plasmin product cleaving the single chain. This two-chain form is regarded as the more active form. Both single chain and two-chain tPA are complexable with PAI-1. PAI-1 acts as an inhibitor for tPA by binding to the tPA and thus stifling its ability to lyse fibrin. tPA can serve as an indicator of both myocardial infarction for patients with impaired fibrinolytic systems as well as a marker for type-II diabetes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty