Would you like to log in again?
Vitamin A & Vitamin E HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
VAE31-H100
102995-084EA 712.69 USD InStock
102995-084
Vitamin A & Vitamin E HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The Vitamin A & Vitamin E HPLC Assay kit determines vitamin A & vitamin E in plasma,serum.
  • Incubation Time: 30 minutes
  • Sample Size: 250 µl
  • Sample Type: Serum, Plasma
Vitamin A and E are fat soluble vitamins. Both are stored in the body, thus overdosing is possible. Vitamin A plays an important role in the visual process. It is mainly taken up as pre vitamin A by nutrition. In the liver it is processed to vitamin A. Retinal is involved in the visual process as prosthetic group of the rhodopsin molecule. Beside this, vitamin A (Retinol) is important for the growth of children and the growth of skin and mucous membranes. As an antioxidant, retinol protects the body against free radicals and reactive oxygen species. Overdosing leads to headache, vertigo, sickness and vomit. Disturbance of the nervous system, skin disease and loss of hair have also been reported.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty