Would you like to log in again?
Vitamin B6 Whole Blood HPLC Assay, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
VB630-H100
102995-090EA 856.54 USD InStock
102995-090
Vitamin B6 Whole Blood HPLC Assay, Eagle Biosciences, Inc.
Assays Immunological Assays
The Vitamin B6 HPLC Assay Kit is intended to detect vitamin B6 in EDTA-blood.
  • Incubation Time: 30 minutes
  • Sample Size: 100 µl
  • Sample Type: EDTA Whole Blood
The vitamins pyridoxin, pyridoxal and pyridoxamin and the appropriate phosphate products are summarized as vitamin B6. All forms can be transformed into the active form pyridoxal-5-phosphate. Vitamin B6 functions as a coenzyme and is essential for more than 50 reactions in the protein, carbohydrate and fatty acid metabolism thereby synthesizing, transforming or degrading amino acids. In protein metabolism vitamin B6 supports the resorption of amino acids and their transport into the cells. Furthermore vitamin B6 contributes to the synthesis of neurotransmitters and amine products (histamine).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty