Would you like to log in again?
25-OH Vitamin D3 HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
VD331-H100
102995-092EA 1235.77 USD InStock
102995-092
25-OH Vitamin D3 HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The 25-OH Vitamin D3 HPLC Assay kit is intended to detect 25-OH Vitamin D3 in plasma and serum.
  • Incubation Time: 15 minutes
  • Sample Size: 400 µl
  • Sample Type: Serum, Plasma
The Eagle Biosciences 25-OH Vitamin D3 HPLC Assay kit makes it possible to determine the vitamin in an easy, fast and precise method. The kit includes all reagents in ready to use form for preparation and separation of the samples with exception of the column (IC3401rp) and the controls (IC3401ko). Vitamin D is one of the most important essential bioregulators of the Ca2+- and phosphate metabolism in higher animals. Together with the peptide hormones parathormone and calcitonin it is responsible to obtain the Ca2+- and phosphate homoeostasis. The most important physiological functions of vitamin D are the stimulation of the intestinal resorption of Ca2+ and phosphate and their incorporation in the bone matrix.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty