Would you like to log in again?
Mouse / Rat 25-OH Vitamin D ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
VID21-K01
102995-106EA 680 USD InStock
102995-106
Mouse / Rat 25-OH Vitamin D ELISA, Eagle Biosciences
Assays ELISAs
Mouse Rat 25-OH Vitamin D ELISA detects total 25-OH Vitamin D in serum and EDTA plasma.
  • Dynamic Range: 9 - 150 ng/mL
  • Sensitivity: 0.95 ng/mL
  • Incubation: 2 hours
The group of compounds referred to as Vitamin D, are actually fat soluble steroidal prehormones. The main forms which occur in the body are Vitamin D2 (ergocalciferol) and Vitamin D3 (cholecalciferol). The active form of these molecules is Dihydroxyvitamin D3 (1, 25(OH)2 D3). Vitamin D3 is formed in the skin by photolysis of 7-dehydrocholesterol by ultraviolet radiation from sunlight. It is transported in blood circulation bound to proteins to the liver where it is hydroxylated. Further hydroxylation occurs in the kidneys to produce the most active form. Vitamin D levels are highest in newborns and decrease exponentially throughout life. Sufficient circulating levels of vitamin D are necessary for healthy bone maintenance and cell metabolism. Recent studies have shown that it may also lower incidents of certain cancers. Insufficient levels of Vitamin D can result in osteoporosis and bone fracture in the elderly, secondary hyperparathyroidism, abnormal cell metabolism and even increased incidents of cancer. Severe deficiency may lead to rickets in children and osteomalacia in adults. Disease associated with Vitamin D deficiency may also include: impaired immunity, increased autoimmunity, myopathy, diabetes mellitus, and an increased risk of colon, breast, and prostate cancers. Abnormally high levels (> 200 ng/ml) of Vitamin D leads to Vitamin D toxicity and may cause hypercalcaemia.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty