Would you like to log in again?
25-OH Vitamin D ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
VID31-K01
102995-108EA 680 USD InStock
102995-108
25-OH Vitamin D ELISA, Eagle Biosciences, Inc.
Assays ELISAs
25-OH Vitamin D ELISA Test detects 25-OH Vitamin D in human serum or plasma samples.
  • Dynamic Range: 4 - 120 ng/mL
  • Sensitivity: 1.9 ng/mL
  • Incubation: 2 hours 45 minutes
The Eagle Biosciences 25-OH Vitamin D ELISA assay kit is designed for the serological determination of the vitamin D concentration in the human organism types of vitamin D that are differentiated are vitamin D2 (ergocalciferol) that is contained in plant food (mushrooms, avocado) and vitamin D3 (cholecalciferol) that is produced from 7-dehydrocholesterol in the skin under ultra-violet irradiation or found in animal food or products (sea fish, egg yolk, butter) [1, 2, 3, 4]. These two forms of vitamin D, which are not yet biologically active, are bound by a protein called VDBP (vitamin D binding protein) in the bloodstream, then metabolised in the liver and converted into 25-OH vitamin D2 (calcidiol) and 25-OH vitamin D3 (calcitriol), respectively, which are storage forms of the vitamin with little activity [1]. In contrast to other commercially available tests, the Eagle Biosciences 25-OH Vitamin D ELISA assay kit uses a newly designed monoclonal antibody which is specific for both vitamin D2 and vitamin D3 at 100% specificity. This is necessary because sometimes vitamin D2 instead of D3 is used in therapy [5, 6, 7].
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty