Would you like to log in again?
Anti-ZnT8 (Zinc Transporter 8) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
3791
102995-120EA 2183.85 USD InStock
102995-120
Anti-ZnT8 (Zinc Transporter 8) ELISA, Eagle Biosciences
Assays ELISAs
Anti-ZNT8 (Zinc Transporter 8) ELISA detects Anti-ZNT8 (Zinc Transporter 8) in serum or plasma.
  • Dynamic Range: 10 - 500 U/ml
  • Sensitivity: 10 U/ml
  • Incubation: overnight
The Eagle Biosciences Anti-ZNT8 (Zinc Transporter 8) ELISA Assay kit is intended for the quantitative determination of Anti-ZNT8 (Zinc Transporter 8) in serum or plasma by enzyme linked immunoassay (ELISA). The Eagle Biosciences Anti-ZNT8 ELISA Assay kit is for research use only and not to be used in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty