Would you like to log in again?
High Sensitive Anti-Tg IgG ELISA Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
ATG31-K01
102995-138EA 568.85 USD InStock
102995-138
High Sensitive Anti-Tg IgG ELISA Kit, Eagle Biosciences, Inc.
Assays ELISAs
Anti-Tg (Thyroglobulin) ELISA is intended to detect Anti-Tg (Thyroglobulin) in serum by ELISA.
  • Dynamic Range: 20 - 1280 U/mL
  • Sensitivity: 1 U/mL
  • Incubation: 2 hours
It is a routine practice of measuring serum autoantibodies to thyroglobulin (Tg) and microsomal (TPO) for aid in detecting and monitoring autoimmune thyroid disease. Serum anti-TPO autoantibody and anti-Tg autoantibody are found to be well- correlating with histological changes in Harshimoto’s thyroiditis. Clinically, positive anti-TPO autoantibody is detected in patients with chronic thyroiditis (70-90%), primary hypothyroidism (~60%), thyrotoxicosis (~50%) and thyroid tumors (~17%), however, anti-Tg autoantibody is mainly identified in patients with Harshimoto’s thyroiditis and Graves’ disease (40-70%).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty