Would you like to log in again?
BOND OPEN CONTAINER 7 ML
Supplier: Leica Microsystems
Bond Open Container^ Bond Open Container 7 mL Novocastra, allow the use of reagents from any source on the Bond system. Refilled until a total of 40 mL has been dispensed from it. Ideal for reagents that are consumed intermittently and have a short shelf life.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOND OPEN CONTAINER 7 ML 102995-938 OP79193 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty