Would you like to log in again?
SensoLyte® Anti-MOG(35-55) IgG Quantitative ELISA Kit (Mouse/Rat), AnaSpec
Supplier: AnaSpec
AS-54465
102997-814EA 850.62 USD InStock
102997-814
SensoLyte® Anti-MOG(35-55) IgG Quantitative ELISA Kit (Mouse/Rat), AnaSpec
Assays Enzyme Assays
MOG peptide fragment (35-55) induces autoantibody production and relapsing-remitting neurological disease causing extensive plaque-like demyelination
The SensoLyte® Anti-MOG (35-55) IgG Quantitative ELISA Kit (mouse/rat) provides a convenient, quantitative assay for detecting anti-MOG (35-55) IgG. This colorimetric assay, useful for determining the amount of anti-MOG (35-55) IgG present, can help provide information on the role anti-MOG (35-55) IgG plays in the development of EAE, an in vivo animal model for MS pathogenesis. The assays are performed in a convenient 96-well (strips format).
Autoantibody response to MOG (35-55) is present in multiple sclerosis (MS) patients and MOG (35-55)-induced experimental autoimmune encephalomyelitis (EAE) mice.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty