Would you like to log in again?
SensoLyte® Anti-Mouse MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
Supplier: AnaSpec
AS-55156
102998-092EA 850.62 USD InStock
102998-092
SensoLyte® Anti-Mouse MOG (1-125) IgG Quantitative ELISA Kit, AnaSpec
Assays Enzyme Assays
This kit is optimized to detect anti-mouse MOG (1-125) IgG. Wells are pre-coated with recombinant mouse MOG (1-125) protein and pre-blocked with BSA. The amount of anti-mouse MOG IgG in serum or cerebrospinal fluid is quantified using ELISA. Ample materials and reagents are provided to perform 96 assays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty