Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Multiple Antigenic Peptide

Supplier: AnaSpec
AS-21006
102998-700EA 432.62 USD InStock
102998-700
Multiple Antigenic Peptide
Proteins and Peptides
Multiple Antigenic Peptides (MAPs) are synthetic peptides with 4 branches harboring 2 Lysines each. The Lys residues are used as a scaffold to support 8 peptides. MAPs are used to increase the mass of the peptide entity when used as antigen in immunizations. They represent an alternative to carrier proteins, such as BSA or KLH.
Sequence:K4K2KA - NH2
MW:985.3 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty