Would you like to log in again?
SensoLyte® Anti-Human MOG (1-125) Human IgG Specific ELISA Kit, AnaSpec
Supplier: AnaSpec
AS-55153-H
103001-242EA 979.23 USD InStock
103001-242
SensoLyte® Anti-Human MOG (1-125) Human IgG Specific ELISA Kit, AnaSpec
Assays Enzyme Assays
This kit is optimized to detect anti-human MOG (1-125) IgG in human samples. Wells are pre-coated with recombinant human MOG (1-125) protein and pre-blocked with proprietary solution. The amount of anti- human MOG IgG in serum or plasma is quantified using ELISA. Human anti-Human MOG (1-125) standard is included. Ample materials and reagents are provided to perform 96 assays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty