Would you like to log in again?
SensoLyte® Anti-PLP (139-151) IgG Quantitative ELISA Kit (Mouse) (Colorimetric), AnaSpec
Supplier: AnaSpec
AS-55524
103001-342EA 850.62 USD InStock
103001-342
SensoLyte® Anti-PLP (139-151) IgG Quantitative ELISA Kit (Mouse) (Colorimetric), AnaSpec
Assays Enzyme Assays
SensoLyte® Anti-PLP (139 - 151) IgG Quantitative ELISA Kit (Mouse) is optimized to detect mouse anti-PLP (139-151) IgG. This kit is useful to researchers who wish to determine the amount of anti-PLP (139-151) antibody present in biological samples, and can help provide information on the role it plays in the development and treatment of EAE, an animal model for MS pathogenesis. Wells are pre-coated with PLP (139-151) peptide and pre-blocked with BSA. The amount of anti-PLP (139-151) IgG in mouse serum or cerebrospinal fluid is quantified using ELISA (Abs=450 nm). Ample materials and reagents are provided to perform 96 assays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty