Would you like to log in again?
Anti-Mouse/ Rat β-Amyloid (1-42) Quantitative ELISA (Colorimetric), AnaSpec
Supplier: AnaSpec
AS-55554
103001-640EA 868.15 USD InStock
103001-640
Anti-Mouse/ Rat β-Amyloid (1-42) Quantitative ELISA (Colorimetric), AnaSpec
Assays Enzyme Assays
This SensoLyte® high-sensitivity (2pg/mL) beta-Amyloid (1-42) Quantitative ELISA Kit (Mouse/Rat) provides a convenient and quantitative assay for determining mouse/rat beta-Amyloid (1-42) (Aβ42) amount in cell and tissue lysate as well as in body fluids
Alzheimer's Disease (AD) is the most common neurodegenerative disorder in elderly people. It has been demonstrated that AD has biological causes and is characterized by the presence of senile plaques and neurofibrillary tangles mainly in cerebral cortex and hippocampus brain regions. Beta-Amyloid (1-40) (Aβ40) and beta-Amyloid (1-42) (Aβ42) are the main components of the above plaques; however, other forms of beta-Amyloid peptides are also present. Both peptides are cleaved from the Amyloid Precursor Protein (APP) by β-secretase and γ-secretase enzymes. Many studies suggest that Aβ42 or/and Aβ43 are required to initiate formation of amyloid plaques and neurofibrills that leads to the neurodegeneration,while Aβ40 is less neurotoxic.
Compared to other mouse/rat anti-Aβ42 ELISA kits on the market, it takes less time to run this assay. HRP conjugated detection antibody in this kit is added simultaneously with the samples and standards during the assay. This eliminates extra incubation and washing steps and makes this kit a one-step procedure for Aβ42 quantification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty