Would you like to log in again?
EndoClear™ Human Recombinant alpha-Synuclein (from E. coli)
Supplier: AnaSpec
EndoClear™
AS-55555-500 AS-55555-100 AS-55555-1000
103001-646EA 584.62 USD InStock
103001-646 103001-644 103001-642
EndoClear™ Human Recombinant alpha-Synuclein (from E. coli)
Proteins and Peptides
Full-length Recombinant human alpha-synuclein (GenBank Accession # NP_000336) was expressed in E. coli., and purified from bacterial lysate using proprietary method. Endotoxin was further removed by proprietary technique.
α-Synuclein is a major component of Lewy bodies in the affected neurons in Parkinson's disease. This protein has a mass of 14.5 kDa (140 amino acids long) and consists of a conserved degenerative amino-terminal domain and an acidic carboxyl-terminal with higher sequence divergence. α-Synuclein is predominantly expressed in brain, specifically in cerebellum, thalamus, neocortex, hippocampus, and striatum regions. Other tissues express α-Synuclein at very low levels. The physiological role of α-synuclein is not yet well understood. However, the presence of imperfect KTKEGV lipid interacting repeats suggests that it may be involved in synaptic vesicle homeostasis.
The human α-synuclein recombinant proteins from AnaSpec are offered with low endotoxin levels of <0.1 EU per μg protein labeled as EndoClear™ products.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty