Would you like to log in again?
Human Recombinant Tau (from E. coli) GST Tag
Supplier: AnaSpec
AS-55557-50 AS-55557-100
103001-726EA 330.31 USD InStock
103001-726 103001-724
Human Recombinant Tau (from E. coli) GST Tag
Proteins and Peptides
The sequence (Accession # AAC04279.1) corresponding to the full length human Tau-441 (2N4R isoform) protein along with GST tag at N-terminal was expressed in E. coli. The recombinant GST-Tau-441 protein was purified from bacterial lysate using proprietary method.
Microtubule associated protein (Tau) is found predominantly in the central neural system and its major function is to promote assembly and to stabilize neuronal microtubules. Six isoforms of Tau were identified in humans that are differentiated by the exclusion or inclusion of exons 2, 3, and 10. Tau-441 is the longest of Tau isoforms, consisting of 441 amino acids with molecular mass of 45.8 kDa. Under physiological conditions Tau can undergo abnormal phosphorylation, truncation, or other modifications that result in the protein detachment from microtubules. These modified Tau molecules can self-associate and form different types of aggregates including neurofibrillary tangles (NFTs) found in brains of patients with neurodegenerative diseases such as Alzheimer’s disease.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty