Would you like to log in again?
Mouse Recombinant MOG (1-125) (from E. coli)
Supplier: AnaSpec
AS-55150-1000 AS-55150-100 AS-55150-500
103004-648EA 1448.38 USD InStock
103004-648 103004-644 103004-646
Mouse Recombinant MOG (1-125) (from E. coli)
Proteins and Peptides
Myelin Oligodendrocyte Glycoprotein (MOG) is a member of the immunoglobulin superfamily and is expressed exclusively in the central nervous system (CNS). Although MOG protein constitutes only 0.01-0.05% of the CNS myelin proteins, it was demonstrated that MOG protein is a crucial autoantigen for multiple sclerosis in humans and experimental autoimmune encephalomyelitis (EAE) in rodents and monkeys
The sequence (Accession # NP_034944) corresponding to the extracellular domain of mouse MOG along with a 6x His tag was expressed in E. coli. The recombinant mouse MOG (M-rMOG) was purified from urea denatured bacterial lysate using immobilized metal affinity chromatography (IMAC). The molecular mass of the recombinant mouse MOG is 14.2 kDa
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty