Would you like to log in again?
Human Recombinant alpha-Synuclein (from E. coli), HiLyte Fluor® 488
Supplier: AnaSpec
AS-55457
103004-984EA 993.85 USD InStock
103004-984
Human Recombinant alpha-Synuclein (from E. coli), HiLyte Fluor® 488
Proteins and Peptides
The recombinant human a-synuclein (1-140) (GenBank Accession # NP_000336) was expressed and purified from E. coli and conjugated with the fluorescence dye HiLyte FluorTM 488. This protein is produced without affinity tag.
α-Synuclein is a major component of Lewy bodies in the affected neurons in Parkinson's disease. This protein has a mass of 14.5 kDa (140 amino acids long) and consists of a conserved degenerative amino-terminal domain and an acidic carboxyl-terminal with higher sequence divergence. α-Synuclein is predominantly expressed in brain, specifically in cerebellum, thalamus, neocortex, hippocampus, and striatum regions. Other tissues express α-Synuclein at very low levels. The physiological role of α-synuclein is not yet well understood. However, the presence of imperfect KTKEGV lipid interacting repeats suggests that it may be involved in synaptic vesicle homeostasis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty