Would you like to log in again?
BOND Novocastra™ Epitope Retrieval Solution, Leica
Supplier: Leica
RE7113-CE RE7119-CE
103006-052EA 737.71 USD InStock
103006-052 102971-662
BOND Novocastra™ Epitope Retrieval Solution, Leica
Media for Microbiology Additives and Supplements for Microbiology
Novocastra™ Epitope Retrieval Solutions are intended for Heat Induced Epitope Retrieval (HIER) on formalin-fixed, paraffin-embedded tissue sections as part of an immunohistochemical procedure
A universal HIER technique suitable for all epitopes does not exist. A combination of different heating methods and epitope retrieval solutions may be used to optimize unmasking of antigens where this technique is recommended. HIER is not recommended for all antibodies.
Delivery information: RE7113 is supplied as a 1 L volume, sufficient to prepare 10 L of working solution. It is a citrate-based buffer and should be diluted with de-ionized water to prepare a working solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty