Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ClearPoint™ BSA peptide

Supplier: AnaSpec
ClearPoint™
AS-61220
103006-434EA 306.92 USD InStock
103006-434
ClearPoint™ BSA peptide
Proteins and Peptides
Mass spec standard
Sequence:DAF-L*-GSF-L*-YEYSR [L* = L(U13C6, 15N)]
MW:1581.7 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty