Would you like to log in again?
SensoLyte® MFP Acid Phosphatase Assay Kit Fluorimetric, AnaSpec
Supplier: AnaSpec
AS-71231
103008-890EA 577.31 USD InStock
103008-890
SensoLyte® MFP Acid Phosphatase Assay Kit Fluorimetric, AnaSpec
Assays Enzyme Assays
The fluorescence of hydrolyzed product of MFP is pH-insensitive, thus it can be used to continuously monitor phosphatase activity in acidic buffer
The change of acid phosphatase level and activity is involved in a variety of physiological and pathological events, such as prostate puberty, rheumatoid arthritis, bone-resorption related diseases, serum marker of tumor bone metastasis, and diabetes. SensoLyte® Acid Phosphatase Assay Kits uses specific substrates to quantify specific acid phosphatase activities such as MFP, FDP or acid phosphatases. The kit can be used for continuously monitoring phosphatase activity in acidic buffer and is compatible with high throughput screening liquid hadnling instruments.
Optimized to detect protein phosphatase activity; compatible with HTS; provides enough reagents to perform 500 assays in a 96-well format; minimal hands-on time; detailed protocol and references provided.
SensoLyte® MFP Acid Phosphatase Assay Kit uses MFP to quantify acid phosphatase activities. The assay is highly sensitive and has large dynamic range.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty