Would you like to log in again?
SensoLyte® 520 TACE ( α-Secretase) Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72085
103008-962EA 850.62 USD InStock
103008-962
SensoLyte® 520 TACE ( α-Secretase) Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
TACE (tumor necrosis factor-alpha converting enzyme), α-secretase, or ADAM17 is a member of the ADAM family of proteases that contain both disintegrin and metalloprotease (catalytic) domains
SensoLyte® 520 TACE Activity Assay Kit uses a 5-FAM (fluorophore) and QXL® 520 (quencher) labeled FRET peptide substrate for continuous measurement of enzyme activity. In the intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage of the FRET peptide by the active enzyme, the fluorescence of 5-FAM is recovered, and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, 5-FAM/QXL® 520 based FRET peptide has less interference from the autofluorescence of test compounds and cellular components and provides better assay sensitivity. The kit can be used to detect the activity of TACE or screen for TACE inhibitors. The assays are performed in a convenient 96-well format.
This enzyme is involved in sperm-egg binding and fusion, muscle cell fusion, neurogenesis, modulation of Notch receptor and ligand processing, and processing of the pro-inflammatory cytokine, TNF α. TACE is currently being explored as a target for anti-inflammatory drugs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty