Would you like to log in again?
SensoLyte® 390 ACE2 Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72086
103008-964EA 787.77 USD InStock
103008-964
SensoLyte® 390 ACE2 Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
ACE2 or ACEH, angiotensin-converting enzyme 2, a homolog of angiotensin-converting enzyme, is a novel zinc metallopeptidase with specificity, tissue distribution, and function distinct from those of ACE
The SensoLyte® 390 ACE2 Activity Assay Kit provides a convenient assay for high throughput screening of ACE2 enzyme inhibitors and inducers using a Mc-Ala/Dnp fluorescence resonance energy transfer (FRET) peptide. In the FRET peptide, the fluorescence of Mc-Ala is quenched by Dnp. Upon cleavage into two separate fragments by ACE2, the fluorescence of Mc-Ala is recovered, and can be monitored at excitation/emission = 330/390 nm. This assay can detect the activity of subnanogram level of ACE2. Assays are performed in a convenient 96-well microplate format.
ACE2 may play a unique role in the renin-angiotensin system and mediate cardiovascular and renal function.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty