Would you like to log in again?
SensoLyte® Green Pin1 Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72240
103008-974EA 931 USD InStock
103008-974
SensoLyte® Green Pin1 Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Peptidyl-prolyl isomerase (PPIase) Pin1 catalyzes cis/trans isomerization of the phospho-Serine/Threonine-Proline peptide bonds
The SensoLyte® Green Pin1 Assay Kit uses a fluorogenic substrate in the cis isoform. Pin1 changes substrate into trans conformation, which is then readily cleaved to generate fluorescent signal which can be monitored at Ex/Em=490/520nm. Increase in fluorescence intensity is directly proportional to the Pin1 activity.
Pin1 consists of short N-terminal protein-protein interaction domain that allows enzyme to bind phosphoproteins and longer C-terminal izomerase domain. Pin1 has many biological substrates, plays critical role in cell-cycle regulation, and up-regulated in many human cancers. Recently, Pin1 was linked to the Alzheimer’s disease pathogenesis. Pin1 can bind phosphorylated Thr-212/231 residues of Tau protein and increase its deposphorylation that may prevent formation of Tau neurofibrilary tangles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty