Would you like to log in again?
SensoLyte® Rh110 Matriptase Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72241
103008-976EA 850.62 USD InStock
103008-976
SensoLyte® Rh110 Matriptase Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Matriptase (also known as MT-SP1, ST14, TADG-15 and epithin) is a trypsin-like protease from the family of type II transmembrane serine proteases
The SensoLyte® Rh110 Matriptase Assay Kit provides a convenient assay for screening of enzyme inhibitors and activators or for continuous assay of enzyme activity using a fluorogenic substrate. Upon matriptase cleavage, this substrate generates the Rh110 (Rhodamine 110) fluorophore which has a bright green fluorescence and can be detected at excitation/emission=496 nm/520 nm. The longer wavelength spectra and higher extinction coefficient of Rh110 provide greater sensitivity and less interference from other reaction components. This assay can detect as low as 0.15 ng/mL active Matriptase.
The spectrum of known Matriptase substrates includes extracellular matrix proteins, cell adhesion molecules, ion channels, growth-factor-like proteins and other proteases. Its actions can result in protein processing, activation or degradation. Matriptase is widely expressed in virtually all epithelium and is specifically found in tumors of epithelial origin. Matriptase has been implicated in carcinogenesis including ovarian, prostate and cervical cancers and is considered a drug target for developing cancer therapeutic agent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty