Would you like to log in again?
SensoLyte® 520 Acetylcholinesterase Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72242
103008-978EA 532 USD InStock
103008-978
SensoLyte® 520 Acetylcholinesterase Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
The SensoLyte® 520 Acetylcholinesterase Assay Kit is a homogeneous assay that can be used to detect the activity of enzyme and for screening of AChE inhibitors
Acetylcholinesterase (AChE) is a fast acting serine hydrolase that belongs to the carboxylesterase enzyme family. It hydrolyses acetylcholine into choline and acetic acid and is found in the synaptic cleft. It can be detected in many mammals and non-mammalian species. Alzheimer and myasthenia gravis diseases are characterized by reduced acetylcholine. Thus, drugs that inhibit AChE were developed to reduce disease symptoms. AChE can be inhibited by many natural substances such as alkaloids and synthetic chemicals such as organophosphorous compounds that are frequently used as pesticides. Recently, it was demonstrated that natural compounds have variable inhibitory effect on AChE depending on the species.
Active AChE generates fluorescence that can be monitored at excitation/emission=490/520 nm. The long wavelength fluorescence of the substrate in this kit minimizes interference from autofluorescence of components in biological samples and test compounds, resulting in higher sensitivity. Optimized to detect human AChE. Detects as low as 8 pg/mL of active AChE. Entire process can be completed in 1 hour.
Caution: For Research Use Only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty