Would you like to log in again?
SensoLyte® 520 ECEs Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72243
103008-980EA 931 USD InStock
103008-980
SensoLyte® 520 ECEs Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
ECEs or Endothelin converting enzymes (ECE-1/ECE-2) are known as rate-limiting enzymes for the formation of the vasoconstrictor peptide endothelin-1 (ET-1) from its precursor proendothelin-1
The SensoLyte® 520 ECEs Activity Assay Kit can be used to detect ECE activity in biological samples or in purified enzyme preparations. The substrate provided in this kit was based on a beta amyloid fragment. When active ECEs cleave the FRET substrate, it results in an increase of 5-FAM fluorescence, and is monitored at excitation/emission = 490/520 nm. The long wavelength fluorescence of 5-FAM shows less interference from autofluorescence of components in biological samples and test compounds. This assay can detect as low as 15.6 ng/mL active ECE-1 and 62.5 ng/mL active ECE-2.
ECE-1 can also degrade amyloid ß fragments both in vitro and in vivo. Highly homologous and structurally comparable to ECE-1, ECE-2 was shown in homozygous knockout mice to have decreased Aß degradation and significant elevated Aß 1-40 and Aß 1-42 in the brain.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty