Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ECAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-13596R-FITC
10301-598EA 811.1 USD InStock
10301-598
Anti-ECAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
ECAT1 (ES cell-associated transcript 1 protein) is a 217 amino acid protein that belongs to the KHDC1 family. The ECAT1 protein contains an atypical KH domain with amino acid changes at critical sites, suggesting that it may not bind RNA. Expression of ECAT1 appears to be maximal in germinal vesicle oocytes, it tails off through metaphase II oocytes and is undetectable following the completion of the oocyte to embryo transition. Specifically expressed in the oocytes, recent studies suggest that ECAT1 may function as a regulator of genomic imprinting in the oocyte. Defects in ECAT1 are the cause of hydatidiform mole recurrent type 2 (HYDM2), a disorder characterized by excessive trophoblast development that produces a growing mass of tissue inside the uterus at the beginning of a pregnancy. HYDM2 leads to abnormal pregnancies with no embryo, and cystic degeneration of the chorionic villi.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ECAT1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty