Would you like to log in again?
SensoLyte® MFP Protein Phosphatase Assay Kit Fluorimetric, AnaSpec
Supplier: AnaSpec
AS-71104
103010-116EA 532 USD InStock
103010-116
SensoLyte® MFP Protein Phosphatase Assay Kit Fluorimetric, AnaSpec
Assays Enzyme Assays
MFP is a proprietary fluorogenic substrate for most phosphatases, alkaline/acid/protein tyrosine/serine/threonine phosphatases
Protein phosphatases have received great attention as drug-screening targets. SensoLyte® Protein Phosphatase Assay Kits uses specific substrates to quantify specific protein phosphatase activities such as MFP, pNPP FDP or acid phosphatases. The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of protein phosphatase inhibitors/inducers.
Optimized to detect protein phosphatase activity; compatible with HTS; provides enough reagents to perform 500 assays in a 96-well format; minimal hands-on time; detailed protocol and references provided.
The phosphatase-induced hydrolysis of MFP yields an intense fluorescent product that has excitation wavelength around 488 nm, and maximum emission around 520 nm. This characteristic makes the substrate adaptable to all the fluorescence instruments. SensoLyte® MFP Protein Phosphatase Assay Kit uses MFP to quantify protein phosphatase activities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty