Would you like to log in again?
SensoLyte® FDP Alkaline Phosphatase Assay Kit Fluorimetric, AnaSpec, Inc.
Supplier: AnaSpec
AS-71109
103010-122EA 507.15 USD InStock
103010-122
SensoLyte® FDP Alkaline Phosphatase Assay Kit Fluorimetric, AnaSpec, Inc.
Assays Enzyme Assays
Conjugates of calf intestinal alkaline phosphatase are extensively used as secondary detection reagents in ELISAs, immunohistochemical techniques and Northern, Southern and Western blot analyses
  • Features: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.
  • Optimized to detect specific alkaline phosphatase activity; compatible for HTS; detailed protocol and references provided.
Phosphatases are important classes of enzymes involved in cellular signal transduction, cell metabolism and other biological activities. Changes in alkaline phosphatase level and activity are involved in a variety of physiological and pathological events, such as bone development, bone-related diseases, gestation related diseases, inflammatory bowel disease, post-parathyroidectomy stage and drug toxicity. Phosphatases have attracted considerable interest as: Enzyme markers, Drug screening targets, Conjugating agents and Reporter for gene expression studies.
The SensoLyte® FDP Alkaline Phosphatase Assay Kit uses highly purified FDP to quantify alkaline phosphatase activity in solutions, in cell extracts, in live cells as well as on solid surfaces (such as PVDF membranes).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty