Would you like to log in again?
SensoLyte® ADHP Peroxidase Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71111
103010-128EA 372.69 USD InStock
103010-128
SensoLyte® ADHP Peroxidase Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
HRP conjugates are extensively used as secondary detection reagents in ELISAs, immuno-histochemical techniques and Northern, Southern and Western blot analyses.
SensoLyte® ADHP Peroxidase Assay Kit uses highly purified ADHP to quantify peroxidase activity in solutions, in cell extracts and in live cells and on solid surfaces (such as PVDF membranes). The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of oxidase inhibitors. The kit inlcludes ADHP peroxidase substrate (Ex/Em=573/590 nm upon oxidation) and an optimized ‘mix and read’ assay protocol that is compatible with HTS liquid handling instruments, among all essential assay components.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty