Would you like to log in again?
SensoLyte® ADHP Hydrogen Peroxide Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71112
103010-130EA 372.69 USD InStock
103010-130
SensoLyte® ADHP Hydrogen Peroxide Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Hydrogen peroxide is involved in a number of biological events, in particular, free radical-induced biochemical reactions.
SensoLyte® ADHP Hydrogen Peroxide Assay Kit uses highly purified ADHP to quantify hydrogen peroxide in solutions, in cell extracts and in live cells. It can also be used to detect a variety of oxidase activities through enzyme-coupled reactions. The kit includes ADHP phosphatase substrate (Ex/Em=573/590 nm upon oxidation) and an optimized ‘mix and read’ assay protocol that is compatible with HTS liquid handling instruments, among other essential assay components.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty