Would you like to log in again?
SensoLyte® Homogeneous AFC Caspase-3/7 Assay Kit, AnaSpec Inc.
Supplier: AnaSpec
AS-71114
103010-132EA 543.69 USD InStock
103010-132
SensoLyte® Homogeneous AFC Caspase-3/7 Assay Kit, AnaSpec Inc.
Assays Enzyme Assays
Central to the execution phase of apoptosis are the two closely related caspase-3 and caspase-7
The SensoLyte® Homogeneous AFC Capase-3/7 Assay Kit uses Ac-DEVD-AFC as the fluorogenic indicator for assaying caspase-3/7 activities. Upon caspase-3/7 cleavage, Ac-DEVD-AFC generates the AFC fluorophore which has bright blue fluorescence and can be detected at Ex/Em=380 nm/500 nm. A bi-function assay buffer in this kit is designed to lyze the cells and measure the enzyme activity at the same time. Thus, this kit can measure caspase-3/7 activity in cell culture directly in a 96-well or 384-well plate without a time-consuming cell extraction step. In case the cells are cultured in larger plates or flasks, a lysis buffer and protocol for cell lysate preparation are also conveniently included in the kit. The kit is suitable for high throughput screening of apoptosis inducers and inhibitors.
They share common substrate specificity and structure, but differ completely in the sequence of their respective N-terminal regions. Both caspase-3 and caspase-7 have substrate selectivity for the amino acid sequence Asp-Glu-Val-Asp (DEVD).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty