Would you like to log in again?
SensoLyte® AFC Caspase Profiling Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71116
103010-134EA 1236.46 USD InStock
103010-134
SensoLyte® AFC Caspase Profiling Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Caspases play important roles in apoptosis and cell signaling
The SensoLyte® Caspase Profiling Kit contains a series of AFC-based peptide substrates (Ex/Em=380 nm/500 nm) as fluorogenic indicators for assaying caspase protease activities. The kit contains a well-designed plate in which a series of AFC-based caspase substrates are coated with both positive and negative controls. It provides the best solution for profiling caspases or caspase inhibitors.
They are also identified as drug-screening targets. AFC-based substrates yield blue fluorescence upon protease cleavage. They are widely used to monitor caspase activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty