Would you like to log in again?
SensoLyte® Green Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71124
103010-144EA 507.15 USD InStock
103010-144
SensoLyte® Green Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
The SensoLyte® Green Protease Assay Kit is widely used for detection of generic protease activities. The kit uses a casein derivative that is heavily labeled with a rhodamine derivative, resulting in almost total quenching of the conjugate's fluorescence. Protease-catalyzed hydrolysis relieves this quenching conjugate, yielding brightly green fluorescent dye-labeled peptides. The increase in fluorescence intensity is directly proportional to protease activity (Ex/Em=488/520 nm). The SensoLyte® protease assay kits do not require any separation steps and can be used to continuously measure the kinetics of a variety of exopeptidases and endopeptidases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty