Would you like to log in again?
SensoLyte® 490 HIV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71127
103010-148EA 1175.08 USD InStock
103010-148
SensoLyte® 490 HIV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
HIV protease (PR) is identified as an important drug-screening target for the design of selective acquired immunodeficiency syndrome (AIDS) therapeutics.
The SensoLyte® 490 HIV Protease Assay Kit uses an optimized FRET peptide substrate for a continuous measurement of HIV protease activities. This FRET-based fluorogenic substrate is derived from a natural processing site for HIV-1 PR. Incubation of the recombinant HIV-1 PR with this substrate results in specific cleavage and a time-dependent increase in fluorescence intensity that is linearly related to the extent of substrate hydrolysis. Because of its simplicity and precision in the determination of reaction rates required for kinetic analysis, this kit offers many advantages over the commonly used HPLC or electrophoresis-based assays for peptide substrate hydrolysis by retroviral PRs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty