Would you like to log in again?
SensoLyte® 490 MMP-1 Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71128
103010-150EA 1175.08 USD InStock
103010-150
SensoLyte® 490 MMP-1 Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
MMP-1 (interstitial collagenase, fibroblast collagenase) is an extracellular protease
The SensoLyte® 490 MMP-1 Assay Kit uses an optimized fluorogenic peptide substrate for continuous measurement of the enzyme activity, providing speed, convenience and accuracy in analyzing MMP-1 and screening MMP-1 inhibitors. Members of the MMP family have poor substrate sequence specificity, making it difficult to use a peptide substrate alone to differentiate the activity of a particular MMP from other MMPs. If several MMPs are coexisting in your samples and you would like to specifically measure MMP-1 activity, please choose the SensoLyte® Plus MMP-1 Assay Kit.
This enzyme plays a significant role in some diseases and has been identified as a drug screening target.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty