Would you like to log in again?
SensoLyte® 490 MMP-2 Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71129
103010-152EA 1175.08 USD InStock
103010-152
SensoLyte® 490 MMP-2 Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® 490 MMP-2 Assay Kit is a complete assay system designed to continuously analyze MMP activities or to screen MMP-2 inhibitors by using a fluorogenic MMP-2 substrate. The assays are performed in a convenient 96-well or 384-well microplate format. The kit provides speed, convenience and accuracy in analyzing MMP-2 activities and in screening MMP-2 inhibitors.
The importance of MMPs in tumor development and invasion is well known. MMP-2 (gelatinase A) is proposed as a potential therapeutic target.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty