Would you like to log in again?
SensoLyte® 490 MMP-9 Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-71134
103010-160EA 1175.08 USD InStock
103010-160
SensoLyte® 490 MMP-9 Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® 490 MMP-9 Assay Kit is a complete assay system designed to continuously analyze MMP activity or to screen for MMP-9 inhibitors using a fluorogenic MMP-9 substrate. The assays are performed in a convenient 96-well or 384-well microplate format. The kit provides speed, convenience and accuracy in MMP-9 analysis. Members of the MMP family have poor substrate sequence specificity, making it difficult to use a peptide substrate alone to differentiate the activity of a particular MMP from other MMPs. If several MMPs are coexisting in your samples and you would like to specifically measure MMP-9 activity, please choose the SensoLyte® Plus MMP-9 Assay Kit.
MMP-9 (gelatinase-B, collagenase-IV) is involved in quite a few diseases such as a cancer, angiogenesis, alopecia and metastasis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty