Would you like to log in again?
SensoLyte® Red Protease Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-71140
103010-168EA 507.15 USD InStock
103010-168
SensoLyte® Red Protease Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
The SensoLyte® Red Protease Assay Kit is widely used for continuous detection of generic protease activities. The kit uses a casein derivative that is heavily labeled with a rhodamine derivative, resulting in almost total quenching of the conjugate's fluorescence. Compared to the SensoLyte® Green Protease Assay Kit, this kit contains a red fluorescent casein derivative. Protease-catalyzed hydrolysis relieves this quenching conjugate, yielding brightly red fluorescent dye-labeled peptides (Ex/Em=546/575 nm). The increase in fluorescence intensity is directly proportional to protease activity. The SensoLyte® protease assay kits do not require any separation steps and can be used to continuously measure the kinetics of a variety of exopeptidases and endopeptidases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty