Would you like to log in again?
SensoLyte® Homogeneous Rh110 Caspase-3/7 Assay Kit
Supplier: AnaSpec
AS-71141
103010-170EA 558.31 USD InStock
103010-170
SensoLyte® Homogeneous Rh110 Caspase-3/7 Assay Kit
Assays Enzyme Assays
Central to the execution phase of apoptosis are the two closely related caspase-3 and caspase-7
The SensoLyte® Homogeneous Rh110 Capase-3/7 Assay Kit uses (Ac-DEVD)2-Rh110 as the fluorogenic indicator for assaying caspase-3/7 activities. Upon caspase-3/7 cleavage, (Ac-DEVD)2-Rh110 generates the Rh110 fluorophore which has bright green fluorescence and can be detected at Ex/Em=496 nm/520 nm. The longer-wavelength spectra and higher extinction coefficient of the Rh110 provide greater sensitivity and less interference from cell components.A bi-function assay buffer in this kit is designed to lyze the cells and measure the enzyme activity at the same time. Thus, this kit can measure caspase-3/7 activity in cell culture directly in a 96-well or 384-well plate without a time-consuming cell extraction step. In case the cells are cultured in larger plates or flasks, a lysis buffer and protocol for cell lysate preparation are also conveniently included in the kit. The kit is suitable for high throughput screening of apoptosis inducers and inhibitors.
They share common substrate specificity and structure, but differ completely in the sequence of their respective N-terminal regions. Both caspase-3 and caspase-7 have substrate selectivity for the amino acid sequence Asp-Glu-Val-Asp (DEVD).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty