Would you like to log in again?
SensoLyte® 520 β - Secretase Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-71144
103010-172EA 868.15 USD InStock
103010-172
SensoLyte® 520 β - Secretase Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
β-Secretase is a membrane-anchored aspartic protease existing in acidic subcellular vesicles
The SensoLyte® 520 β-Secretase Assay Kit is a homogeneous assay that can be used to detect the activity of enzyme and for screening of secretase inhibitors. It contains a QXL® 520/ HiLyte™ Fluor 488 FRET substrate. The sequence of this FRET peptide is derived from the β-secretase cleavage site on the Swedish APP mutation. This mutation causes the enhancement of β-secretase to process APP, resulting in an early onset of AD. Active β-secretase cleaves FRET substrate into two separate fragments resulting in the release of HiLyte™ Fluor 488 fluorescence that can be monitored at excitation/emission= 490/520 nm. The long wavelength fluorescence of HiLyte™ Fluor 488 is less interfered by the autofluorescence of components in biological samples and test compounds.
It is a key player in producing β-amyloid peptides through proteolytic cleavage of the β-amyloid precursor protein (APP). Accumulation of neurotoxic β-amyloid peptide is seen in senile plaques in the brains of patients with Alzheimer’s disease (AD), an age-related cognitive disorder. Therefore, targeting β-secretase is considered a potential therapeutic strategy for the prevention and treatment of AD.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty