Would you like to log in again?
SensoLyte® 520 HCV Protease Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-71145
103010-174EA 850.62 USD InStock
103010-174
SensoLyte® 520 HCV Protease Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
The SensoLyte® 520 HCV Assay Kit uses a new FRET peptide substrate that incorporates 5-FAM (fluorophore) and QXL® 520 (quencher) for a continuous measurement of enzyme activities. In the intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage of the FRET peptide by HCV NS3/4a protease, the fluorescence of 5-FAM is recovered and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, this 5-FAM/QXL® 520-based FRET peptide shows less interference from autofluorescence of test compounds and cellular components. This assay is ten times more sensitive than an EDANS/DABCYL based assay and can detect 0.1 pmole of HCV NS3/4a protease. The assays are performed in a convenient 96-well or 384-well microplate format.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty