Would you like to log in again?
AnaTag™ B-PE Labeling Kit, Anaspec Inc.
Supplier: AnaSpec
AS-72112
103010-182EA 925.15 USD InStock
103010-182
AnaTag™ B-PE Labeling Kit, Anaspec Inc.
Protein Linking and Labeling Systems
The AnaTag™ B-PE Labeling Kit is optimized for use in the conjugation of B-Phycoerythrin (B-PE) to antibodies. B-PE, a fluorescent protein from phycobiliprotein family, has a primary absorption peak at 545 nm with a secondary peak at 563 nm and maximum emission at 578 nm.
B-PE and the closely related R-PE are the most intensely fluorescent phycobiliproteins having orange fluorescence. They are significantly brighter and more photostable than conventional organic fluorophores. B-PE labeled primary and secondary antibodies are used in applications such as flow cytometry and multi-color immunofluorescent staining. The AnaTagTM B-PE Labeling Kit contains SH-reactive B-PE. SMCC modified B-PE reacts with the thiol groups of target antibody without the need for additional activation, thus simplifying the conjugation protocol. The amount of B-PE supplied in this kit is sufficient for labeling up to 1 mg of antibody. The kit provides all reagents, purification columns and a detailed step-by-step protocol. Native and SMCC-activated B-PE are also available as separate products.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty