Would you like to log in again?
AnaTag™ R-PE Labeling Kit, AnaSpec Inc.
Supplier: AnaSpec
AS-72113
103010-184EA 925.15 USD InStock
103010-184
AnaTag™ R-PE Labeling Kit, AnaSpec Inc.
Assays Enzyme Assays
The AnaTag™ R-PE Labeling Kit is optimized for use in the conjugation of R-Phycoerythrin (R-PE) to antibodies. R-PE, a fluorescent protein from phycobiliprotein family, has broad absorption bands with peaks at 565 nm (eM =1.96 x 106 M-1cm-1), 498 (eM =1.53 x 106 M-1cm-1), and 539 nm (eM =1.62 x 106 M-1cm-1); it therefore can be excited with versatile excitation sources. Its maximal emission can be detected at 578 nm. The broad excitation spectrum also provides the advantage of multi-color immunofluorescent staining or cell sorting. R-PE and the closely related B-PE are the most intensely fluorescent phycobiliproteins having orange fluorescence. They are significantly brighter and more photostable than conventional organic fluorophores. The AnaTagTM R-PE Labeling Kit contains SH-reactive R-PE. SMCC modified R-PE reacts with the thiol groups of target antibody without the need for additional activation, thus simplifying the conjugation protocol. The amount of R-PE supplied in this kit is sufficient for labeling up to 1 mg of antibody. The kit provides all reagents, purification columns and a detailed step-by-step protocol. Native and SMCC-activated R-PE are also available as separate products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty