Would you like to log in again?
SensoLyte® 520 MMP Substrate Sampler Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-71170
103010-248EA 1236.46 USD InStock
103010-248
SensoLyte® 520 MMP Substrate Sampler Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
SensoLyte® 520 MMP Substrate Sampler Kit contains a series of FRET peptide substrates for use as fluorogenic indicators to assay MMP protease activities. It provides the best solution for identifying a suitable fluorogenic substrate for a particular MMP assay. All the MMP substrates in the kit are also available in bulk packages.
They are involved in diseases such as cancer, arthritis, asthma, angiogenesis, alopecia and metastasis. They are also identified as drug-screening targets.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty